Assay Details
Target Gene Details
Entrez Gene ID: | 3696 |
Gene Name: | integrin subunit beta 8 |
Gene Aliases: |
- |
Location: |
Chr.7:20329819-20415759 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGB8 | NM_002214.2 | NP_002205.1 | ||
XM_011515393.2 | XP_011513695.1 | |||
XM_011515394.2 | XP_011513696.1 | |||
XM_011515396.1 | XP_011513698.1 | |||
XM_017012178.1 | XP_016867667.1 | |||
XM_017012179.1 | XP_016867668.1 | |||
XM_017012180.1 | XP_016867669.1 | |||
XM_017012181.1 | XP_016867670.1 | |||
XM_017012182.1 | XP_016867671.1 | |||
XM_017012183.1 | XP_016867672.1 | |||
AK295044.1 | ||||
BC002630.2 | AAH02630.1 | |||
BI861225.1 | ||||
BT006739.1 | AAP35385.1 | |||
M73780.1 | AAA36034.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)