Assay Details
Target Gene Details
Entrez Gene ID: | 2050 |
Gene Name: | EPH receptor B4 |
Gene Aliases: |
HTK, MYK1, TYRO11 |
Location: |
Chr.7:100802565-100827521 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB4 | NM_004444.4 | NP_004435.3 | ||
XM_017011816.1 | XP_016867305.1 | |||
AB209644.1 | BAD92881.1 | |||
AK301224.1 | ||||
AY056048.1 | AAL14195.1 | |||
BC004264.1 | AAH04264.1 | |||
BC052804.1 | AAH52804.1 | |||
U07695.1 | AAA20598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759547 | Chr.7:100757867 - 101105965 on Build GRCh38 | Gain+Loss | MUC17 MUC12 SLC12A9 LOC102724094 ACHE MIR6875 UFSP1 SRRT ZAN EPHB4 TRIM56 TRIP6 MUC3A |
nsv470381 | Chr.7:100642568 - 100834358 on Build GRCh38 | Loss | POP7 GNB2 EPO GIGYF1 ACTL6B TFR2 LOC105375429 ZAN EPHB4 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | AZGP1P1 MIR25 STAG3L5P-PVRIG2P-PILRB GAL3ST4 ZASP ACTL6B SPDYE3 LOC102724094 PVRIG2P VGF PILRB SRRT ZAN PMS2P1 LRCH4 TRIM56 LAMTOR4 TSC22D4 PPP1R35 MBLAC1 TRIP6 C7orf43 GATS POP7 GIGYF1 ZSCAN21 STAG3L5P AZGP1 ACHE ZKSCAN1 TAF6 PVRIG PCOLCE CNPY4 AP1S1 MUC17 ZNHIT1 MUC12 SLC12A9 PLOD3 MIR4658 ZCWPW1 FIS1 C7orf61 PCOLCE-AS1 MIR106B MEPCE EPHB4 STAG3 NAT16 MOGAT3 PILRA AP4M1 GPC2 MIR6840 SAP25 GNB2 TFR2 MIR4653 MOSPD3 MCM7 MIR6875 UFSP1 SERPINE1 ZNF3 COPS6 CLDN15 EPO FBXO24 LOC105375429 NYAP1 MUC3A AGFG2 MIR93 |
nsv607965 | Chr.7:100776465 - 100899511 on Build GRCh38 | Loss | SLC12A9 ACHE MIR6875 UFSP1 SRRT ZAN TRIP6 EPHB4 |
nsv1020837 | Chr.7:100715499 - 100911015 on Build GRCh38 | Loss | EPO SLC12A9 ACHE MIR6875 UFSP1 SRRT ZAN TRIP6 EPHB4 |
nsv951388 | Chr.7:100741378 - 100838178 on Build GRCh38 | Deletion | ZAN EPHB4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |