Assay Details
Target Gene Details
Entrez Gene ID: | 404744 |
Gene Name: | NPSR1 antisense RNA 1 |
Gene Aliases: |
AAA1 |
Location: |
Chr.7:34346512-34834331 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPSR1-AS1 | NR_033664.1 | |||
NR_033665.1 | ||||
AY312368.1 | AAQ85136.1 | |||
AY312369.1 | AAQ85137.1 | |||
AY312370.1 | AAQ85138.1 | |||
AY321516.1 | AAQ87610.1 | |||
BX649082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion | NPSR1-AS1 ZNRF2P1 LOC100421336 LINC00997 KBTBD2 DPY19L1P2 MIR548N RP9 DPY19L1P1 RP9P MIR550B2 BMPER NT5C3A FKBP9 MIR550A2 DPY19L1 NPSR1 FLJ20712 BBS9 |
esv3612792 | Chr.7:34177815 - 34353101 on Build GRCh38 | Gain | NPSR1-AS1 |
esv3891097 | Chr.7:33915559 - 34483233 on Build GRCh38 | Gain | NPSR1-AS1 BMPER |
esv3612772 | Chr.7:33730657 - 34432276 on Build GRCh38 | Loss | NPSR1-AS1 BMPER |
nsv1024493 | Chr.7:33733357 - 34425137 on Build GRCh38 | Loss | NPSR1-AS1 BMPER |
nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion | RP9P NPSR1-AS1 BMPER NT5C3A LOC100421336 FKBP9 KBTBD2 NPSR1 FLJ20712 DPY19L1P2 RP9 BBS9 |
nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion | NPSR1-AS1 ZNRF2P1 LOC100421336 LINC00997 KBTBD2 DPY19L1P2 MIR548N RP9 DPY19L1P1 RP9P MIR550B2 BMPER NT5C3A FKBP9 MIR550A2 DPY19L1 NPSR1 FLJ20712 BBS9 |