Assay Details
Target Gene Details
Entrez Gene ID: | 404744 |
Gene Name: | NPSR1 antisense RNA 1 |
Gene Aliases: |
AAA1 |
Location: |
Chr.7:34346512-34834331 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPSR1-AS1 | NR_033664.1 | 5 | 4284 | |
NR_033665.1 | 4 | 4134 | ||
BX649082.1 | 5 | 4247 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3612792 | Chr.7:34177815 - 34353101 on Build GRCh38 | Gain |
![]() |
esv3891097 | Chr.7:33915559 - 34483233 on Build GRCh38 | Gain |
![]() ![]() |
esv3612772 | Chr.7:33730657 - 34432276 on Build GRCh38 | Loss |
![]() ![]() |
nsv1024493 | Chr.7:33733357 - 34425137 on Build GRCh38 | Loss |
![]() ![]() |
nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)