Assay Details
Target Gene Details
Entrez Gene ID: | 81552 |
Gene Name: | vesicular, overexpressed in cancer, prosurvival protein 1 |
Gene Aliases: |
ECOP, GASP |
Location: |
Chr.7:55434406-55572520 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VOPP1 | XM_011515539.1 | XP_011513841.1 | ||
XM_011515540.2 | XP_011513842.1 | |||
XM_011515541.2 | XP_011513843.1 | |||
XM_011515546.2 | XP_011513848.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2734425 | Chr.7:53868335 - 55917254 on Build GRCh38 | Deletion | VOPP1 HPVC1 EGFR-AS1 EGFR LOC107986797 VSTM2A-OT1 LOC100996654 LOC101060341 ELDR LINC01445 VSTM2A SEC61G FKBP9P1 LANCL2 SEPT14 ZNF713 PSPHP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |