Assay Details
Target Gene Details
Entrez Gene ID: | 23072 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
NEDL1 |
Location: |
Chr.7:43112580-43566001 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
Target Gene Details
Entrez Gene ID: | 100506895 |
Gene Name: | uncharacterized LOC100506895 |
Gene Aliases: |
- |
Location: |
Chr.7:43508728-43522542 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100506895 | NR_038276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3612993 | Chr.7:43438543 - 43609526 on Build GRCh38 | Gain | STK17A LOC100506895 HECW1 COA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |