Assay Details
Target Gene Details
Entrez Gene ID: | 6677 |
Gene Name: | sperm adhesion molecule 1 |
Gene Aliases: |
HEL-S-96n, HYA1, HYAL1, HYAL3, HYAL5, PH-20, PH20, SPAG15 |
Location: |
Chr.7:123925232-123971410 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPAM1 | NM_003117.4 | NP_003108.2 | ||
BC026163.1 | AAH26163.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |