Assay Details
Target Gene Details
Entrez Gene ID: | 23072 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
NEDL1 |
Location: |
Chr.7:43112580-43566001 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HECW1 | NM_001287059.1 | NP_001273988.1 | ||
NM_015052.4 | NP_055867.3 | |||
XM_005249665.3 | XP_005249722.1 | |||
XM_006715670.3 | XP_006715733.1 | |||
XM_006715671.3 | XP_006715734.1 | |||
XM_006715673.3 | XP_006715736.1 | |||
XM_017011882.1 | XP_016867371.1 | |||
XM_017011883.1 | XP_016867372.1 | |||
XM_017011884.1 | XP_016867373.1 | |||
XM_017011885.1 | XP_016867374.1 | |||
XM_017011886.1 | XP_016867375.1 | |||
XM_017011887.1 | XP_016867376.1 | |||
XM_017011888.1 | XP_016867377.1 | |||
XM_017011889.1 | XP_016867378.1 | |||
XM_017011890.1 | XP_016867379.1 | |||
AB002320.2 | ||||
AK090799.1 | ||||
AK294918.1 | ||||
BC151227.1 |
Target Gene Details
Entrez Gene ID: | 100127950 |
Gene Name: | HECW1 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.7:43117896-43163187 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HECW1-IT1 | NR_135295.1 | |||
AK093262.1 | BAC04113.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |