Assay Details
Target Gene Details
Entrez Gene ID: | 9723 |
Gene Name: | semaphorin 3E |
Gene Aliases: |
M-SEMAH, M-SemaK, SEMAH, coll-5 |
Location: |
Chr.7:83363906-83649163 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEMA3E | NM_001178129.1 | NP_001171600.1 | ||
NM_012431.2 | NP_036563.1 | |||
AB002329.1 | ||||
AK297209.1 | ||||
AK303925.1 | ||||
BC140706.1 | ||||
BC144338.1 | ||||
BC152458.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1023612 | Chr.7:82592183 - 83790140 on Build GRCh38 | Loss | SEMA3E PCLO |
esv3613933 | Chr.7:83362430 - 83451993 on Build GRCh38 | Loss | SEMA3E |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |