Assay Details
Target Gene Details
Entrez Gene ID: | 89845 |
Gene Name: | ATP binding cassette subfamily C member 10 |
Gene Aliases: |
EST182763, MRP7, SIMRP7 |
Location: |
Chr.6:43427538-43451994 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABCC10 | NM_001198934.1 | NP_001185863.1 | ||
XM_011514974.2 | XP_011513276.2 | |||
XM_011514983.2 | XP_011513285.1 | |||
XM_017011445.1 | XP_016866934.1 | |||
XM_017011446.1 | XP_016866935.1 | |||
XM_017011447.1 | XP_016866936.1 | |||
XM_017011448.1 | XP_016866937.1 | |||
XM_017011449.1 | XP_016866938.1 | |||
XM_017011450.1 | XP_016866939.1 | |||
XM_017011451.1 | XP_016866940.1 | |||
XM_017011452.1 | XP_016866941.1 | |||
XM_017011453.1 | XP_016866942.1 | |||
XM_017011454.1 | XP_016866943.1 | |||
XM_017011455.1 | XP_016866944.1 | |||
AK000002.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv603000 | Chr.6:43397622 - 43524840 on Build GRCh38 | Loss | MIR6780B ABCC10 DLK2 XPO5 LRRC73 YIPF3 TJAP1 POLR1C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |