Assay Details
Target Gene Details
Entrez Gene ID: | 9841 |
Gene Name: | zinc finger and BTB domain containing 24 |
Gene Aliases: |
BIF1, ICF2, PATZ2, ZNF450 |
Location: |
Chr.6:109462516-109483237 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZBTB24 | NM_014797.2 | NP_055612.2 | ||
AB007901.1 | ||||
AK309697.1 | ||||
BC117374.1 | ||||
BC117376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1153690 | Chr.6:109469796 - 109472297 on Build GRCh38 | Duplication |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)