Assay Details
Target Gene Details
Entrez Gene ID: | 2295 |
Gene Name: | forkhead box F2 |
Gene Aliases: |
FKHL6, FREAC-2, FREAC2 |
Location: |
Chr.6:1389834-1395597 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXF2 | NM_001452.1 | NP_001443.1 | ||
U13220.1 | AAC32226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950756 | Chr.6:1380966 - 1396765 on Build GRCh38 | Deletion | FOXF2 LOC105374880 MIR6720 |
nsv1017614 | Chr.6:1248415 - 1423267 on Build GRCh38 | Loss | LOC107986554 FOXF2 LOC105374880 MIR6720 FOXQ1 LINC01394 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |