Assay Details
Target Gene Details
Entrez Gene ID: | 51534 |
Gene Name: | vesicle trafficking 1 |
Gene Aliases: |
C6orf55, DRG-1, DRG1, HSPC228, LIP5, My012, SBP1 |
Location: |
Chr.6:142147162-142220948 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VTA1 | NM_001286371.1 | NP_001273300.1 | ||
NM_001286372.1 | NP_001273301.1 | |||
NM_016485.4 | NP_057569.2 | |||
AF060225.1 | AAG43125.1 | |||
AF141341.1 | AAG49444.1 | |||
AF151062.1 | AAF36148.1 | |||
AF271994.1 | AAF76210.1 | |||
AJ420507.1 | ||||
AK000051.1 | BAA90909.1 | |||
AK130738.1 | ||||
AK301376.1 | ||||
AL136684.1 | CAB66619.1 | |||
BC005937.1 | AAH05937.1 | |||
BC006989.1 | AAH06989.1 | |||
BC022536.1 | AAH22536.1 | |||
BM806661.1 | ||||
CR457207.1 | CAG33488.1 | |||
DA299959.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830824 | Chr.6:142039970 - 142223792 on Build GRCh38 | Gain | NMBR VTA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |