Assay Details
Target Gene Details
Entrez Gene ID: | 7957 |
Gene Name: | epilepsy, progressive myoclonus type 2A, Lafora disease (laforin) |
Gene Aliases: |
EPM2, MELF |
Location: |
Chr.6:145500744-145736018 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPM2A | XM_011536113.2 | XP_011534415.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)