Assay Details
Target Gene Details
Entrez Gene ID: | 7468 |
Gene Name: | Wolf-Hirschhorn syndrome candidate 1 |
Gene Aliases: |
MMSET, NSD2, REIIBP, TRX5, WHS |
Location: |
Chr.4:1871357-1982207 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WHSC1 | NM_001042424.2 | NP_001035889.1 | ||
NM_007331.1 | NP_015627.1 | |||
NM_133330.2 | NP_579877.1 | |||
NM_133331.2 | NP_579878.1 | |||
NM_133334.2 | NP_579889.1 | |||
NM_133335.3 | NP_579890.1 | |||
XM_005248001.3 | XP_005248058.1 | |||
XM_005248002.2 | XP_005248059.1 | |||
XM_005248005.2 | XP_005248062.1 | |||
XM_006713914.3 | XP_006713977.1 | |||
XM_011513557.2 | XP_011511859.1 | |||
AF071593.1 | AAC24150.1 | |||
AF071594.1 | AAC24151.1 | |||
AF083386.1 | AAD19343.1 | |||
AF083387.1 | AAD21770.1 | |||
AF083388.1 | AAD21771.1 | |||
AF083389.1 | AAD19344.1 | |||
AF083390.1 | AAD19345.1 | |||
AF083391.1 | AAD19346.1 | |||
AJ007042.1 | CAB45386.1 | |||
AK289697.1 | ||||
AY694128.1 | AAU09264.1 | |||
BC020545.1 | ||||
BC052254.1 | AAH52254.1 | |||
BC141815.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |