Assay Details
Target Gene Details
Entrez Gene ID: | 10608 |
Gene Name: | MAX dimerization protein 4 |
Gene Aliases: |
MAD4, MST149, MSTP149, bHLHc12 |
Location: |
Chr.4:2247432-2262143 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MXD4 | NM_006454.2 | NP_006445.1 | ||
XM_017007655.1 | XP_016863144.1 | |||
XM_017007656.1 | XP_016863145.1 | |||
AF040963.1 | AAB97009.1 | |||
AF082740.1 | AAF98553.1 | |||
AF187724.1 | AAQ13695.1 | |||
BC002713.2 | AAH02713.1 | |||
BC068060.1 | AAH68060.1 | |||
BT019969.1 | AAV38772.1 | |||
BT019970.1 | AAV38773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508990 | Chr.4:2246239 - 2324327 on Build GRCh38 | Insertion | MIR4800 ZFYVE28 MXD4 |
nsv593377 | Chr.4:2246106 - 2261919 on Build GRCh38 | Loss | MIR4800 MXD4 |
nsv4200 | Chr.4:2252081 - 2286377 on Build GRCh38 | Insertion | ZFYVE28 MXD4 |
nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain | RNF4 POLN MIR4800 HAUS3 ZFYVE28 MXD4 CFAP99 |
nsv1073719 | Chr.4:2260372 - 2262373 on Build GRCh38 | Deletion | MXD4 |
dgv8912n54 | Chr.4:2257123 - 2262637 on Build GRCh38 | Loss | MXD4 |
esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion | MIR4800 HAUS3 TNIP2 CFAP99 NOP14-AS1 C4orf48 ADD1 NAT8L RNF4 POLN ZFYVE28 FAM193A MXD4 NOP14 SH3BP2 MFSD10 |
nsv593388 | Chr.4:2257730 - 2266399 on Build GRCh38 | Gain | MXD4 |
nsv527189 | Chr.4:2243138 - 2276689 on Build GRCh38 | Loss | MIR4800 ZFYVE28 MXD4 |
dgv8911n54 | Chr.4:2253738 - 2262338 on Build GRCh38 | Loss | MXD4 |
dgv8910n54 | Chr.4:2249015 - 2262496 on Build GRCh38 | Loss | MIR4800 MXD4 |
nsv829838 | Chr.4:2155167 - 2349950 on Build GRCh38 | Loss | POLN MIR4800 HAUS3 ZFYVE28 MXD4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |