Assay Details
Target Gene Details
Entrez Gene ID: | 285521 |
Gene Name: | COX18, cytochrome c oxidase assembly factor |
Gene Aliases: |
COX18HS |
Location: |
Chr.4:73054696-73069759 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX18 | NM_001297732.1 | NP_001284661.1 | ||
NM_001297733.1 | NP_001284662.1 | |||
NM_001300729.1 | NP_001287658.1 | |||
NM_173827.3 | NP_776188.1 | |||
XM_005265680.4 | XP_005265737.1 | |||
XM_011531878.2 | XP_011530180.1 | |||
XM_017008045.1 | XP_016863534.1 | |||
AK096310.2 | BAC04758.1 | |||
AM055750.1 | CAJ20055.1 | |||
AM055751.1 | CAJ20056.1 | |||
AM055752.1 | CAJ20057.1 | |||
AY957564.1 | AAY35060.1 | |||
AY957565.1 | AAY35061.1 | |||
BC101684.1 | AAI01685.1 | |||
BC143642.1 | ||||
DA385228.1 | ||||
DA674605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3600985 | Chr.4:72952867 - 73192514 on Build GRCh38 | Gain |
![]() ![]() |
nsv523169 | Chr.4:72886525 - 73255193 on Build GRCh38 | Gain |
![]() ![]() |
esv3600984 | Chr.4:72944988 - 73087723 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)