Assay Details
Target Gene Details
Entrez Gene ID: | 1259 |
Gene Name: | cyclic nucleotide gated channel alpha 1 |
Gene Aliases: |
CNCG, CNCG1, CNG-1, CNG1, RCNC1, RCNCa, RCNCalpha, RP49 |
Location: |
Chr.4:47935015-48016718 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNGA1 | NM_000087.3 | NP_000078.2 | ||
NM_001142564.1 | NP_001136036.1 | |||
XM_005248049.4 | XP_005248106.2 | |||
XM_011513623.2 | XP_011511925.1 | |||
XM_017007712.1 | XP_016863201.1 | |||
AK292021.1 | ||||
M84741.1 | AAA52010.1 | |||
S42457.1 | AAB22778.1 | |||
X64802.1 | CAA46027.1 |
Target Gene Details
Entrez Gene ID: | 101927157 |
Gene Name: | uncharacterized LOC101927157 |
Gene Aliases: |
- |
Location: |
Chr.4:47914227-47990737 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927157 | NR_125879.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |