Assay Details
Target Gene Details
Entrez Gene ID: | 441024 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like |
Gene Aliases: |
- |
Location: |
Chr.4:74123280-74303099 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTHFD2L | XM_017008225.1 | XP_016863714.1 | ||
XM_017008226.1 | XP_016863715.1 | |||
XM_017008231.1 | XP_016863720.1 | |||
XM_017008232.1 | XP_016863721.1 | |||
BC032771.1 | AAH32771.1 | |||
BG566009.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain | CXCL5 PF4 CXCL3 PPBP CXCL2 PPBPP2 MTHFD2L |
nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain | LOC105377276 CXCL5 PF4 CXCL1 CXCL3 EPGN EREG PPBP AREG CXCL2 PPBPP2 MTHFD2L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |