Assay Details
Target Gene Details
Entrez Gene ID: | 132724 |
Gene Name: | transmembrane protease, serine 11B |
Gene Aliases: |
- |
Location: |
Chr.4:68226653-68245694 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS11B | NM_182502.3 | NP_872308.2 | ||
XM_011531608.2 | XP_011529910.1 | |||
XM_011531609.1 | XP_011529911.1 | |||
AK290904.1 | ||||
AK310210.1 | ||||
AL833167.1 | CAD91168.1 | |||
BC126195.1 | ||||
BX537945.1 | CAD97913.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv594506 | Chr.4:68183925 - 68357339 on Build GRCh38 | Loss | TMPRSS11B TMPRSS11BNL YTHDC1 FTLP10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |