Assay Details
Target Gene Details
Entrez Gene ID: | 101410542 |
Gene Name: | UCHL1 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:41220074-41256727 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UCHL1-AS1 | NR_102709.1 | |||
CN277289.1 | ||||
DB169394.1 |