Assay Details
Target Gene Details
Entrez Gene ID: | 64151 |
Gene Name: | non-SMC condensin I complex subunit G |
Gene Aliases: |
CAPG, CHCG, NY-MEL-3, YCG1 |
Location: |
Chr.4:17810813-17844865 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCAPG | NM_022346.4 | NP_071741.2 | ||
NR_073124.1 | ||||
XM_011513876.2 | XP_011512178.1 | |||
XM_017008543.1 | XP_016864032.1 | |||
XM_017008544.1 | XP_016864033.1 | |||
AB013299.1 | BAB21557.1 | |||
AF235023.1 | AAG30732.1 | |||
AF331796.1 | AAG49627.1 | |||
AK022512.1 | BAB14069.1 | |||
AK023147.1 | BAB14429.1 | |||
BC068467.1 | AAH68467.1 | |||
BC101476.1 | AAI01477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893889 | Chr.4:17807915 - 17817512 on Build GRCh38 | Loss | NCAPG DCAF16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |