Assay Details
Target Gene Details
Entrez Gene ID: | 100505875 |
Gene Name: | long intergenic non-protein coding RNA 1088 |
Gene Aliases: |
- |
Location: |
Chr.4:78971748-79308799 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01088 | NR_038342.1 |
Target Gene Details
Entrez Gene ID: | 84779 |
Gene Name: | N(alpha)-acetyltransferase 11, NatA catalytic subunit |
Gene Aliases: |
ARD1B, ARD2, hARD2 |
Location: |
Chr.4:79155237-79326058 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA11 | XM_017008779.1 | XP_016864268.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1014428 | Chr.4:79168558 - 79211659 on Build GRCh38 | Gain | NAA11 LINC01088 |
esv2727859 | Chr.4:78716995 - 79557290 on Build GRCh38 | Deletion | LOC101928893 GK2 NAA11 LINC00989 BMP2K MIR5096 LINC01088 PAQR3 |
nsv1006026 | Chr.4:78111837 - 79966019 on Build GRCh38 | Loss | LOC101928893 LINC00989 ANXA3 ANTXR2 MIR5096 LINC01094 LINC01088 PAQR3 FRAS1 GK2 NAA11 BMP2K PCAT4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |