Assay Details
Target Gene Details
Entrez Gene ID: | 658 |
Gene Name: | bone morphogenetic protein receptor type 1B |
Gene Aliases: |
ALK-6, ALK6, AMDD, BDA1D, BDA2, CDw293 |
Location: |
Chr.4:94757977-95158453 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BMPR1B | NM_001203.2 | NP_001194.1 | ||
XM_011532201.2 | XP_011530503.1 | |||
XM_017008559.1 | XP_016864048.1 | |||
AW299723.1 | ||||
U89326.1 | AAC28131.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)