Assay Details
Target Gene Details
Entrez Gene ID: | 101927179 |
Gene Name: | uncharacterized LOC101927179 |
Gene Aliases: |
- |
Location: |
Chr.4:47831345-47896848 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927179 | NR_125880.1 | |||
NR_125881.1 | ||||
AL599247.1 | ||||
BX280044.1 |
Target Gene Details
Entrez Gene ID: | 152518 |
Gene Name: | nuclear transcription factor, X-box binding like 1 |
Gene Aliases: |
CDZFP, HOZFP, OZFP, URCC5 |
Location: |
Chr.4:47847233-47914667 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFXL1 | NM_001278623.1 | NP_001265552.1 | ||
NM_001278624.1 | NP_001265553.1 | |||
NM_152995.5 | NP_694540.3 | |||
NR_103795.1 | ||||
AB085695.1 | ||||
AB181916.1 | ||||
AK131285.1 | BAD18459.1 | |||
AK302440.1 | ||||
AY134856.1 | AAN08626.1 | |||
BC018019.1 | AAH18019.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5202n100 | Chr.4:47486414 - 47904818 on Build GRCh38 | Gain | MIR8053 NFXL1 LOC101927179 ATP10D CORIN |
nsv4328 | Chr.4:47814450 - 47860236 on Build GRCh38 | Deletion | NFXL1 LOC101927179 CORIN |
nsv594118 | Chr.4:47558369 - 47915713 on Build GRCh38 | Loss | MIR8053 NFXL1 LOC101927179 ATP10D CORIN LOC101927157 |
nsv829924 | Chr.4:47668888 - 47864648 on Build GRCh38 | Gain | NFXL1 LOC101927179 CORIN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |