Assay Details
Target Gene Details
Entrez Gene ID: | 9348 |
Gene Name: | N-deacetylase and N-sulfotransferase 3 |
Gene Aliases: |
HSST3 |
Location: |
Chr.4:118033596-118258945 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDST3 | NM_004784.2 | NP_004775.1 | ||
XM_006714416.3 | XP_006714479.1 | |||
XM_011532415.2 | XP_011530717.1 | |||
XM_011532416.2 | XP_011530718.1 | |||
XM_017008839.1 | XP_016864328.1 | |||
XM_017008840.1 | XP_016864329.1 | |||
AF074924.1 | AAD15978.1 | |||
AK295439.1 | ||||
AY358852.1 | AAQ89211.1 | |||
BC109309.1 | AAI09310.1 | |||
BC109310.1 | AAI09311.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759279 | Chr.4:117557693 - 118753587 on Build GRCh38 | Gain | LOC101929741 LOC729218 METTL14 SEC24D SNHG8 SNORA24 NDST3 CEP170P1 PRSS12 LINC01378 |
dgv5372n100 | Chr.4:118028335 - 118350088 on Build GRCh38 | Loss | SNHG8 SNORA24 NDST3 PRSS12 |
nsv519017 | Chr.4:117907652 - 118850326 on Build GRCh38 | Loss | LOC101929741 LOC729218 METTL14 SEC24D SNHG8 SNORA24 NDST3 CEP170P1 PRSS12 |
esv34354 | Chr.4:117643184 - 118666956 on Build GRCh38 | Gain | LOC101929741 LOC729218 SNHG8 SNORA24 NDST3 CEP170P1 PRSS12 LINC01378 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |