Assay Details
Target Gene Details
Entrez Gene ID: | 4547 |
Gene Name: | microsomal triglyceride transfer protein |
Gene Aliases: |
ABL, MTP |
Location: |
Chr.4:99564078-99623997 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTTP | NM_000253.3 | NP_000244.2 | ||
NM_001300785.1 | NP_001287714.1 | |||
AK302602.1 | ||||
DB083001.1 | ||||
DB458800.2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |