Assay Details
Target Gene Details
Entrez Gene ID: | 131 |
Gene Name: | alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide |
Gene Aliases: |
ADH4 |
Location: |
Chr.4:99412261-99435510 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADH7 | NM_000673.4 | NP_000664.2 | ||
NM_001166504.1 | NP_001159976.1 | |||
AK223472.1 | ||||
AK301657.1 | ||||
AK301696.1 | ||||
AK312854.1 | ||||
BC131512.1 | ||||
CD014081.1 | ||||
L33179.1 | AAA59211.1 | |||
U07821.1 | AAA19002.1 | |||
U09623.1 | AAA82165.1 | |||
X76342.1 | CAA53960.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517344 | Chr.4:99401288 - 99442412 on Build GRCh38 | Gain | ADH7 |
dgv5345n100 | Chr.4:99181544 - 99523288 on Build GRCh38 | Gain | ADH1B ADH7 ADH1A ADH6 ADH1C C4orf17 LOC100507053 |
esv3584519 | Chr.4:99402713 - 99556305 on Build GRCh38 | Gain | TRMT10A ADH7 C4orf17 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |