Assay Details
Target Gene Details
Entrez Gene ID: | 80333 |
Gene Name: | potassium voltage-gated channel interacting protein 4 |
Gene Aliases: |
CALP, KCHIP4 |
Location: |
Chr.4:20728520-21948801 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829876 | Chr.4:20637113 - 20833998 on Build GRCh38 | Gain | PACRGL KCNIP4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |