Assay Details
Target Gene Details
Entrez Gene ID: | 80306 |
Gene Name: | mediator complex subunit 28 |
Gene Aliases: |
1500003D12Rik, EG1, magicin |
Location: |
Chr.4:17614628-17625628 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED28 | NM_025205.4 | NP_079481.2 | ||
AF317678.1 | ||||
AF317679.1 | ||||
AF317680.1 | AAK11563.1 | |||
AF318059.1 | AAL26906.1 | |||
AF358829.1 | AAK32724.1 | |||
AK313928.1 | ||||
BC011936.2 | AAH11936.1 | |||
HY174042.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)