Assay Details
Target Gene Details
Entrez Gene ID: | 51365 |
Gene Name: | phospholipase A1 member A |
Gene Aliases: |
PS-PLA1, PSPLA1 |
Location: |
Chr.3:119597848-119629811 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA1A | NM_001206960.1 | NP_001193889.1 | ||
NM_001206961.1 | NP_001193890.1 | |||
NM_001293225.1 | NP_001280154.1 | |||
NM_015900.3 | NP_056984.1 | |||
NR_120610.1 | ||||
XM_017006572.1 | XP_016862061.1 | |||
AF035268.1 | AAC98921.1 | |||
AF035269.1 | AAC98922.1 | |||
AK222705.1 | BAD96425.1 | |||
AK301880.1 | ||||
AK313519.1 | ||||
BC047703.2 | AAH47703.1 | |||
BX647516.1 | ||||
U37591.1 | AAC99994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv591363 | Chr.3:119597981 - 119627103 on Build GRCh38 | Loss | PLA1A |
nsv829695 | Chr.3:119460043 - 119649149 on Build GRCh38 | Loss | TIMMDC1 POGLUT1 TMEM39A CD80 ADPRH POPDC2 PLA1A |
nsv3963 | Chr.3:119582217 - 119627057 on Build GRCh38 | Deletion | ADPRH PLA1A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |