Assay Details
Target Gene Details
Entrez Gene ID: | 51738 |
Gene Name: | ghrelin/obestatin prepropeptide |
Gene Aliases: |
MTLRP |
Location: |
Chr.3:10285750-10292947 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHRL | NM_001134941.2 | NP_001128413.1 | ||
NM_001134944.1 | NP_001128416.1 | |||
NM_001134945.1 | NP_001128417.1 | |||
NM_001134946.1 | NP_001128418.1 | |||
NM_001302821.1 | NP_001289750.1 | |||
NM_001302822.1 | NP_001289751.1 | |||
NM_001302823.1 | NP_001289752.1 | |||
NM_001302824.1 | NP_001289753.1 | |||
NM_001302825.1 | NP_001289754.1 | |||
NM_016362.4 | NP_057446.1 | |||
NR_126505.1 | ||||
EF549557.1 | ||||
EF549561.1 | ||||
EF549564.1 | ||||
EF549566.1 | ||||
EF549571.1 | ||||
EF549572.1 | ||||
EF549573.1 | ||||
EF549574.1 | ||||
EF549575.1 | ||||
EU072083.1 | ||||
EU072084.1 | ||||
EU072085.1 | ||||
EU072086.1 | ||||
EU072087.1 |
Target Gene Details
Entrez Gene ID: | 100126793 |
Gene Name: | ghrelin opposite strand/antisense RNA |
Gene Aliases: |
GHRL-AS1, GHRLAS, NCRNA00068 |
Location: |
Chr.3:10280952-10293449 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHRLOS | NR_004431.3 | 5 | 1017 | |
NR_024144.2 | 6 | 845 | ||
NR_024145.2 | 6 | 1189 | ||
NR_073566.1 | 7 | 1200 | ||
NR_073567.1 | 6 | 1188 | ||
NR_073568.1 | 6 | 1043 | ||
BX118778.1 | 1 | 178 | ||
EF549568.1 | 4 | 960 | ||
EF549570.1 | 4 | 705 | ||
EU072081.1 | 3 | 892 | ||
EU072082.1 | 2 | 734 | ||
EU789531.1 | 3 | 511 | ||
EU789533.1 | 1 | 59 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | MIR378B ATP2B2 SLC6A11 SEC13 VHL ATG7 MIR885 BRK1 IRAK2 SLC6A1 FANCD2OS ATP2B2-IT2 TAMM41 SLC6A1-AS1 GHRLOS GHRL LINC00852 LOC101927467 VGLL4 LINC00606 TATDN2 FANCD2 HRH1 |
nsv508903 | Chr.3:10275353 - 10401894 on Build GRCh38 | Insertion | MIR378B ATP2B2 SEC13 GHRLOS GHRL LINC00852 MIR885 TATDN2 |
nsv589650 | Chr.3:10260488 - 10294102 on Build GRCh38 | Loss | GHRLOS GHRL LINC00852 TATDN2 |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | MIR378B CIDECP ATG7 MIR885 BRK1 IRAK2 SLC6A1 FANCD2P2 SLC6A1-AS1 LOC401052 LOC101927467 VGLL4 EMC3 LINC00606 TATDN2 FANCD2 ATP2B2 SLC6A11 SEC13 VHL FANCD2OS ATP2B2-IT2 TAMM41 GHRLOS GHRL LINC00852 HRH1 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | MIR378B ATP2B2 SLC6A11 SEC13 VHL ATG7 MIR885 BRK1 IRAK2 SLC6A1 FANCD2OS ATP2B2-IT2 TAMM41 FANCD2P2 SLC6A1-AS1 GHRLOS GHRL LINC00852 LOC101927467 VGLL4 LINC00606 TATDN2 FANCD2 HRH1 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | MIR378B ATP2B2 SLC6A11 SEC13 VHL ATG7 MIR885 BRK1 IRAK2 SLC6A1 FANCD2OS ATP2B2-IT2 TAMM41 FANCD2P2 SLC6A1-AS1 GHRLOS GHRL LINC00852 LOC101927467 VGLL4 LINC00606 TATDN2 FANCD2 HRH1 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | MIR378B CIDECP ATG7 MIR885 BRK1 IRAK2 SLC6A1 FANCD2P2 SLC6A1-AS1 LOC401052 EMC3-AS1 LOC101927467 VGLL4 EMC3 LINC00606 TATDN2 FANCD2 ATP2B2 SLC6A11 SEC13 VHL FANCD2OS ATP2B2-IT2 TAMM41 GHRLOS GHRL LINC00852 HRH1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |