Assay Details
Target Gene Details
Entrez Gene ID: | 79885 |
Gene Name: | histone deacetylase 11 |
Gene Aliases: |
HD11 |
Location: |
Chr.3:13480171-13506424 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HDAC11 | NM_001136041.2 | NP_001129513.1 | ||
NM_024827.3 | NP_079103.2 | |||
XM_005265491.1 | XP_005265548.1 | |||
XM_005265492.1 | XP_005265549.1 | |||
XM_006713338.1 | XP_006713401.1 | |||
XM_011534131.1 | XP_011532433.1 | |||
XM_011534132.2 | XP_011532434.2 | |||
XM_011534135.1 | XP_011532437.1 | |||
XM_017007234.1 | XP_016862723.1 | |||
AK025426.1 | BAB15127.1 | |||
AK025890.1 | BAB15272.1 | |||
AK293223.1 | ||||
AK299021.1 | ||||
AL834223.1 | CAH56375.1 | |||
BC009676.1 | AAH09676.1 | |||
BG708402.1 | ||||
DB025431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470416 | Chr.3:13492082 - 13575549 on Build GRCh38 | Loss | FBLN2 HDAC11 |
nsv1011766 | Chr.3:13485961 - 14213016 on Build GRCh38 | Gain | FGD5P1 TMEM43 LINC00620 XPC WNT7A FBLN2 CHCHD4 LSM3 HDAC11 LOC100293612 TPRXL SNORA93 |
nsv955316 | Chr.3:13491101 - 13497900 on Build GRCh38 | Deletion | HDAC11 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |