Assay Details
Target Gene Details
Entrez Gene ID: | 84100 |
Gene Name: | ADP ribosylation factor like GTPase 6 |
Gene Aliases: |
BBS3, RP55 |
Location: |
Chr.3:97762875-97812585 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARL6 | NR_103511.1 | |||
XM_006713783.3 | XP_006713846.1 | |||
XM_017007311.1 | XP_016862800.1 | |||
XM_017007312.1 | XP_016862801.1 | |||
AK292958.1 | ||||
AK311281.1 | ||||
AL136815.1 | CAB66749.1 | |||
BC024239.2 | AAH24239.1 | |||
BU607306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011526 | Chr.3:97483978 - 97905097 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv520553 | Chr.3:95905623 - 98010484 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1013060 | Chr.3:96856275 - 98493673 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)