Assay Details
Target Gene Details
Entrez Gene ID: | 165829 |
Gene Name: | G protein-coupled receptor 156 |
Gene Aliases: |
GABABL, PGR28 |
Location: |
Chr.3:120165481-120285122 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR156 | NM_001168271.1 | NP_001161743.1 | ||
NM_153002.2 | NP_694547.2 | |||
XM_005247165.2 | XP_005247222.1 | |||
XM_011512484.2 | XP_011510786.1 | |||
XM_011512487.1 | XP_011510789.1 | |||
XM_017005795.1 | XP_016861284.1 | |||
XM_017005796.1 | XP_016861285.1 | |||
XM_017005797.1 | XP_016861286.1 | |||
AB209362.1 | BAD92599.1 | |||
AF488739.1 | AAN03795.1 | |||
BC113701.1 | ||||
BC143606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1013788 | Chr.3:120147875 - 120193450 on Build GRCh38 | Gain |
![]() |
nsv829697 | Chr.3:120084009 - 120199588 on Build GRCh38 | Loss |
![]() ![]() |
nsv3967 | Chr.3:120138861 - 120184268 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)