Assay Details
Target Gene Details
Entrez Gene ID: | 8850 |
Gene Name: | lysine acetyltransferase 2B |
Gene Aliases: |
CAF, P/CAF, PCAF |
Location: |
Chr.3:20040032-20154404 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KAT2B | NM_003884.4 | NP_003875.3 | ||
XM_005265528.4 | XP_005265585.1 | |||
BC060823.1 | AAH60823.1 | |||
BC070075.1 | AAH70075.1 | |||
U57317.2 | AAC50890.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893669 | Chr.3:20001342 - 20074585 on Build GRCh38 | Loss | PP2D1 KAT2B |
esv3893667 | Chr.3:19470582 - 20214665 on Build GRCh38 | Gain | SGO1 EFHB SGO1-AS1 MIR3135A PP2D1 KCNH8 KAT2B RAB5A |
nsv997486 | Chr.3:19965210 - 20072497 on Build GRCh38 | Gain | PP2D1 KAT2B RAB5A |
esv35078 | Chr.3:19513779 - 20728244 on Build GRCh38 | Gain | SGO1 EFHB LOC101927829 SGO1-AS1 MIR3135A PP2D1 KCNH8 KAT2B RAB5A |
esv23068 | Chr.3:19494161 - 20597009 on Build GRCh38 | Gain+Loss | SGO1 EFHB LOC101927829 SGO1-AS1 MIR3135A PP2D1 KCNH8 KAT2B RAB5A |
nsv834627 | Chr.3:20026623 - 20186483 on Build GRCh38 | Loss | SGO1 SGO1-AS1 MIR3135A KAT2B |
nsv433359 | Chr.3:20049416 - 20546080 on Build GRCh38 | Gain | SGO1 LOC101927829 SGO1-AS1 MIR3135A KAT2B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |