Assay Details
Target Gene Details
Entrez Gene ID: | 10093 |
Gene Name: | actin related protein 2/3 complex subunit 4 |
Gene Aliases: |
ARC20, P20-ARC |
Location: |
Chr.3:9792495-9807105 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARPC4 | NM_001024959.2 | NP_001020130.1 | ||
NM_001024960.2 | NP_001020131.1 | |||
NM_001198780.1 | NP_001185709.1 | |||
NM_005718.4 | NP_005709.1 | |||
AF006087.1 | AAB64192.1 | |||
AW408422.1 | ||||
BG938423.1 | ||||
BI768875.1 | ||||
BI910730.1 | ||||
BM045872.1 | ||||
BQ688329.1 | ||||
BX419672.2 | ||||
DA226584.1 | ||||
DB060690.1 |
Target Gene Details
Entrez Gene ID: | 100526693 |
Gene Name: | ARPC4-TTLL3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.3:9792548-9836356 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARPC4-TTLL3 | NM_001198793.1 | NP_001185722.1 | ||
AK024110.1 | BAB14828.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1010768 | Chr.3:9502870 - 9873902 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)