Assay Details
Target Gene Details
Entrez Gene ID: | 152110 |
Gene Name: | NIMA related kinase 10 |
Gene Aliases: |
- |
Location: |
Chr.3:27107336-27369460 on Build GRCh38 |
Assay Gene Location: | Within Intron 42 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv880e214 | Chr.3:26983690 - 27243989 on Build GRCh38 | Gain |
![]() |
esv3595659 | Chr.3:27121167 - 27127959 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)