Assay Details
Target Gene Details
Entrez Gene ID: | 654502 |
Gene Name: | IQ motif containing J |
Gene Aliases: |
- |
Location: |
Chr.3:159069252-159266307 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IQCJ | NM_001042705.2 | NP_001036170.1 | ||
NM_001042706.2 | NP_001036171.1 | |||
NM_001197100.1 | NP_001184029.1 | |||
AK309049.1 | ||||
BC137464.1 | ||||
BC137469.1 | ||||
BC144658.1 | ||||
DA305468.1 | ||||
DQ309553.1 | ||||
DQ309554.1 |
Target Gene Details
Entrez Gene ID: | 100505385 |
Gene Name: | IQCJ-SCHIP1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.3:159069252-159897366 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IQCJ-SCHIP1 | NM_001197113.1 | NP_001184042.1 | ||
NM_001197114.1 | NP_001184043.1 | |||
AK096479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470971 | Chr.3:158873139 - 159440428 on Build GRCh38 | Gain | LOC105374181 IQCJ-SCHIP1 IQCJ SCHIP1 MIR3919 |
nsv1010336 | Chr.3:159009762 - 159147855 on Build GRCh38 | Loss | IQCJ-SCHIP1 IQCJ |
nsv1010828 | Chr.3:159084685 - 159176530 on Build GRCh38 | Gain | IQCJ-SCHIP1 IQCJ |
nsv998949 | Chr.3:159057017 - 159676761 on Build GRCh38 | Gain | IQCJ-SCHIP1 IQCJ SCHIP1 MIR3919 |
esv3893792 | Chr.3:159110854 - 159167887 on Build GRCh38 | Loss | IQCJ-SCHIP1 IQCJ |
esv3598351 | Chr.3:159072467 - 159149169 on Build GRCh38 | Loss | IQCJ-SCHIP1 IQCJ |