Assay Details
Target Gene Details
Entrez Gene ID: | 116832 |
Gene Name: | ribosomal protein L39 like |
Gene Aliases: |
RPL39L1 |
Location: |
Chr.3:187120953-187139475 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPL39L | NM_052969.1 | NP_443201.1 | ||
AB063610.1 | BAC19837.1 | |||
BC012328.1 | AAH12328.1 | |||
L05096.1 | AAC15859.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4157 | Chr.3:187094850 - 187129650 on Build GRCh38 | Insertion |
![]() |
nsv1004008 | Chr.3:186879814 - 187204053 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1014596 | Chr.3:187106902 - 187410668 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)