Assay Details
Target Gene Details
Entrez Gene ID: | 101929152 |
Gene Name: | P3H2 antisense RNA 1 |
Gene Aliases: |
LEPREL1-AS1 |
Location: |
Chr.3:190120964-190144846 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P3H2-AS1 | NR_126419.1 | |||
BM017065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1014822 | Chr.3:190065059 - 190464556 on Build GRCh38 | Gain | P3H2-AS1 CLDN16 CLDN1 TMEM207 P3H2 |
esv3893843 | Chr.3:190075998 - 191094501 on Build GRCh38 | Gain | GMNC P3H2-AS1 SNAR-I CLDN16 CLDN1 TMEM207 IL1RAP P3H2 |