Assay Details
Target Gene Details
Entrez Gene ID: | 25924 |
Gene Name: | myosin VIIA and Rab interacting protein |
Gene Aliases: |
SLAC2-C, SLAC2C |
Location: |
Chr.3:39808914-40260321 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYRIP | NM_001284423.1 | NP_001271352.1 | ||
NM_001284424.1 | NP_001271353.1 | |||
NM_001284425.1 | NP_001271354.1 | |||
NM_015460.3 | NP_056275.2 | |||
NR_104316.1 | ||||
XM_011533575.1 | XP_011531877.1 | |||
AF396687.1 | AAM43954.1 | |||
AK126013.1 | ||||
AK310972.1 | ||||
AK312412.1 | ||||
BC092512.1 | AAH92512.1 | |||
BC109311.1 | AAI09312.1 | |||
BC109312.1 | AAI09313.1 | |||
CF138044.1 | ||||
DN999201.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3773 | Chr.3:39868257 - 39901670 on Build GRCh38 | Insertion |
![]() |
nsv517803 | Chr.3:39859533 - 39991467 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)