Assay Details
Target Gene Details
Entrez Gene ID: | 10954 |
Gene Name: | protein disulfide isomerase family A member 5 |
Gene Aliases: |
PDIR |
Location: |
Chr.3:123067009-123162106 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDIA5 | NM_006810.3 | NP_006801.1 | ||
NR_028444.1 | ||||
BC001625.1 | AAH01625.1 | |||
BM549365.1 | ||||
D49490.1 | BAA08451.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)