Assay Details
Target Gene Details
Entrez Gene ID: | 84126 |
Gene Name: | ATR interacting protein |
Gene Aliases: |
- |
Location: |
Chr.3:48446710-48465655 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATRIP | NM_001271022.1 | NP_001257951.1 | ||
NM_001271023.1 | NP_001257952.1 | |||
NM_032166.3 | NP_115542.2 | |||
NM_130384.2 | NP_569055.1 | |||
AF451323.1 | AAL38042.1 | |||
AK291829.1 | ||||
AK300548.1 | ||||
AK315075.1 | ||||
BC014153.2 | AAH14153.2 | |||
BC020563.2 | AAH20563.1 | |||
BC030597.2 | AAH30597.1 | |||
DC402185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834684 | Chr.3:48344209 - 48551890 on Build GRCh38 | Gain | CCDC51 FBXW12 PFKFB4 TMA7 TREX1 MIR6823 SHISA5 PLXNB1 ATRIP |
nsv834683 | Chr.3:48260014 - 48451855 on Build GRCh38 | Gain | FCF1P2 CCDC51 ZNF589 NME6 FBXW12 TMA7 MIR2115 PLXNB1 SPINK8 ATRIP |
nsv590242 | Chr.3:48392157 - 48518906 on Build GRCh38 | Loss | CCDC51 FBXW12 PFKFB4 TMA7 TREX1 SHISA5 PLXNB1 ATRIP |
nsv508917 | Chr.3:48400224 - 48499357 on Build GRCh38 | Insertion | CCDC51 TMA7 TREX1 SHISA5 PLXNB1 ATRIP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |