Assay Details
Target Gene Details
Entrez Gene ID: | 7018 |
Gene Name: | transferrin |
Gene Aliases: |
HEL-S-71p, PRO1557, PRO2086, TFQTL1 |
Location: |
Chr.3:133661998-133779006 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TF | XM_017007089.1 | XP_016862578.1 | ||
XM_017007090.1 | XP_016862579.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |