Assay Details
Target Gene Details
Entrez Gene ID: | 339896 |
Gene Name: | glutamate decarboxylase like 1 |
Gene Aliases: |
ADC, CSADC, HuADC, HuCSADC |
Location: |
Chr.3:30726009-30894661 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GADL1 | NM_207359.2 | NP_997242.2 | ||
XM_017006297.1 | XP_016861786.1 | |||
XM_017006298.1 | XP_016861787.1 | |||
AK128643.1 | BAC87546.1 | |||
BC093701.1 | AAH93701.1 | |||
BC111986.1 | AAI11987.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)