Assay Details
Target Gene Details
Entrez Gene ID: | 10533 |
Gene Name: | autophagy related 7 |
Gene Aliases: |
APG7-LIKE, APG7L, GSA7 |
Location: |
Chr.3:11272324-11564704 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | GHRLOS HRH1 ATP2B2-IT2 LINC00852 IRAK2 MIR885 MIR378B VHL SLC6A1 ATG7 BRK1 SLC6A11 SEC13 LOC101927467 VGLL4 GHRL TATDN2 ATP2B2 LINC00606 TAMM41 FANCD2OS FANCD2 SLC6A1-AS1 |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | GHRLOS HRH1 LINC00852 IRAK2 MIR885 MIR378B VHL SLC6A1 ATG7 SLC6A11 TATDN2 ATP2B2 FANCD2OS ATP2B2-IT2 CIDECP LOC401052 BRK1 SEC13 LOC101927467 VGLL4 GHRL EMC3 LINC00606 TAMM41 FANCD2 SLC6A1-AS1 FANCD2P2 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | GHRLOS HRH1 ATP2B2-IT2 LINC00852 IRAK2 MIR885 MIR378B VHL SLC6A1 ATG7 BRK1 SLC6A11 SEC13 LOC101927467 VGLL4 GHRL TATDN2 ATP2B2 LINC00606 TAMM41 FANCD2OS FANCD2 SLC6A1-AS1 FANCD2P2 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | GHRLOS HRH1 ATP2B2-IT2 LINC00852 IRAK2 MIR885 MIR378B VHL SLC6A1 ATG7 BRK1 SLC6A11 SEC13 LOC101927467 VGLL4 GHRL TATDN2 ATP2B2 LINC00606 TAMM41 FANCD2OS FANCD2 SLC6A1-AS1 FANCD2P2 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | GHRLOS HRH1 LINC00852 IRAK2 MIR885 MIR378B VHL SLC6A1 ATG7 SLC6A11 TATDN2 ATP2B2 EMC3-AS1 FANCD2OS ATP2B2-IT2 CIDECP LOC401052 BRK1 SEC13 LOC101927467 VGLL4 GHRL EMC3 LINC00606 TAMM41 FANCD2 SLC6A1-AS1 FANCD2P2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |