Assay Details
Target Gene Details
Entrez Gene ID: | 3680 |
Gene Name: | integrin subunit alpha 9 |
Gene Aliases: |
ALPHA-RLC, ITGA4L, RLC |
Location: |
Chr.3:37452322-37819790 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGA9 | NM_002207.2 | NP_002198.2 | ||
BC030198.1 | AAH30198.1 | |||
D25303.1 | BAA04984.1 | |||
L24158.1 | AAA16099.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3595886 | Chr.3:37316808 - 37528893 on Build GRCh38 | Gain | C3orf35 ITGA9 GOLGA4 |
esv3595887 | Chr.3:37319289 - 37878005 on Build GRCh38 | Gain | CTDSPL C3orf35 ITGA9 ITGA9-AS1 GOLGA4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |