Assay Details
Target Gene Details
Entrez Gene ID: | 7325 |
Gene Name: | ubiquitin conjugating enzyme E2 E2 |
Gene Aliases: |
UBCH8 |
Location: |
Chr.3:23203007-23590805 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2E2 | NM_152653.3 | NP_689866.1 | ||
XM_005265432.2 | XP_005265489.1 | |||
XM_005265433.4 | XP_005265490.1 | |||
XM_011534075.1 | XP_011532377.1 | |||
XM_011534076.2 | XP_011532378.1 | |||
XM_017007126.1 | XP_016862615.1 | |||
AK057886.1 | BAB71605.1 | |||
AK307075.1 | ||||
BC022332.1 | AAH22332.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834636 | Chr.3:23097162 - 23259952 on Build GRCh38 | Gain+Loss |
![]() ![]() |
nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)