Assay Details
Target Gene Details
Entrez Gene ID: | 55207 |
Gene Name: | ADP ribosylation factor like GTPase 8B |
Gene Aliases: |
ARL10C, Gie1 |
Location: |
Chr.3:5122245-5180916 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARL8B | NM_018184.2 | NP_060654.1 | ||
AB118751.1 | BAD23992.1 | |||
AK001564.1 | BAA91759.1 | |||
AK295465.1 | ||||
AK298951.1 | ||||
AK303872.1 | ||||
BC013131.1 | AAH13131.1 | |||
BC063125.1 | AAH63125.1 | |||
CR457264.1 | CAG33545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002376 | Chr.3:5146106 - 5235477 on Build GRCh38 | Gain |
![]() ![]() |
esv3595124 | Chr.3:5138599 - 5176477 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)